missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Fluoride Hydrate 98.0+%, TCI America™

Catalog No. p-7136455
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08385G 5 g
T083825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08385G Supplier TCI America Supplier No. T08385G
Only null left

Chemical Identifiers

CAS 665-46-3
Molecular Formula C8H20FN
Molecular Weight (g/mol) 149.253
MDL Number MFCD00011826
InChI Key QSUJAUYJBJRLKV-UHFFFAOYSA-M
PubChem CID 69582
IUPAC Name tetraethylazanium;fluoride
SMILES CC[N+](CC)(CC)CC.[F-]

Specifications

Color White
Quantity 5 g
Formula Weight 149.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium Fluoride Hydrate
EINECSNumber (2)-0186
TSCA Yes