missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Chloride, Monohydrate, Crystal, 98%, Spectrum™ Chemical

Catalog No. 18614350
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614350 25 g Poly Bottle
18614351 100 g Poly Bottle
18614352 500 g Poly Bottle
18614353 1 kg Poly Bottle
18614354 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614350 Supplier Spectrum Chemical Mfg Cor Supplier No. T106625GM

Chemical Identifiers

CAS 68696-18-4

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 25 g
Molecular Weight (g/mol) 183.72
Percent Purity 0.98
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material Tetraethylammonium Chloride, Monohydrate

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.