missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium chloride, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Visit our Sensor Applications portal to learn more.

$197.28 - $935.01

Chemical Identifiers

MDL Number MFCD00011828
Synonym TEA chloride
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111014631
View Documents
Merck Emd Millipore
866165G
5 g
Each for $197.28
Only null left
 
111014630
View Documents
Merck Emd Millipore
8661625G
25 g
Each for $935.01
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00011828
TEA chloride

Specifications

56-34-8
MFCD00011828
TEA chloride
165.70
For electrochemical analysis
(C2H5)4 N(Cl)
5 g
Methanol: 0.1 g/mL, clear, colorless
≥99.0% (AT); ≥99.0%
Videos
Safety and Handling

Safety and Handling

RTECSNumber : BS6125000

SDS
missing translation for 'documents'

missing translation for 'documents'