missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium chloride, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
Supplier: Merck Emd Millipore 8661625G
Specifications
| 56-34-8 | |
| MFCD00011828 | |
| TEA chloride | |
| 165.70 | |
| For electrochemical analysis |
| (C2H5)4 N(Cl) | |
| 25 g | |
| Methanol: 0.1 g/mL, clear, colorless | |
| ≥99.0% (AT); ≥99.0% |
Chemical Identifiers
| MFCD00011828 | |
| TEA chloride |
Safety and Handling
RTECSNumber : BS6125000