missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Bromide in Glacial Acetic Acid, Reagents

Catalog No. 01333464
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333464 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333464 Supplier Labchem Inc Supplier No. CT006600500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetraethylammonium Bromide in Glacial Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-19-7 , 71-91-0
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Tan/Gold
pH <2
Boiling Point 118°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 1.1027
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetraethylammonium Bromide in Glacial Acetic Acid