missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl Propane-1,1,2,3-tetracarboxylate 97.0+%, TCI America™

Catalog No. P048725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P048725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P048725G Supplier TCI America Supplier No. P048725G
Only null left

Chemical Identifiers

CAS 635-03-0
Molecular Formula C15H24O8
Molecular Weight (g/mol) 332.35
MDL Number MFCD00027834
InChI Key WGEVJRBJWMGTSI-UHFFFAOYNA-N
Synonym Propane-1,1,2,3-tetracarboxylic Acid Tetraethyl Ester
PubChem CID 69454
IUPAC Name 1,1,2,3-tetraethyl propane-1,1,2,3-tetracarboxylate
SMILES CCOC(=O)CC(C(C(=O)OCC)C(=O)OCC)C(=O)OCC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 332.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethyl Propane-1,1,2,3-tetracarboxylate