missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl p-Xylylenediphosphonate 95.0+%, TCI America™

Catalog No. T15825G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15825G 5 g
T158225G 25 g
T1582100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T15825G Supplier TCI America Supplier No. T15825G

Chemical Identifiers

CAS 4546-04-7
Molecular Formula C16H28O6P2
Molecular Weight (g/mol) 378.34
MDL Number MFCD00671576
InChI Key XTKQUBKFKSHRPS-UHFFFAOYSA-N
Synonym p-Bis(diethylphosphono)xylene, p-Xylylenediphosphonic Acid Tetraethyl Ester
PubChem CID 78303
IUPAC Name 1,4-bis(diethoxyphosphorylmethyl)benzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)CP(=O)(OCC)OCC)OCC

Specifications

Melting Point 76°C
Color White
Boiling Point 220°C
Quantity 5 g
Formula Weight 378.34
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethyl p-Xylylenediphosphonate