accessibility menu, dialog, popup

UserName

H3COPOPOOCH3OCH3OCH3

CAS RN 4546-04-7

IUPAC Name: diethyl ({4-[(diethoxyphosphoryl)methyl]phenyl}methyl)phosphonate
Synonyms: diethyl {4-[(diethoxyphosphoryl)methyl]phenyl}methylphosphonate
Molecular Weight (g/mol): 378.34
Molecular Formula: C16H28O6P2
InChi Key: XTKQUBKFKSHRPS-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(CP(=O)(OCC)OCC)C=C1)OCC
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results