missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyltrimethylammonium Bromide 98.0+%, TCI America™

Catalog No. p-7136490
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T090625G 25 g
T0906100G 100 g
T0906500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T090625G Supplier TCI America Supplier No. T090625G
Only null left

Chemical Identifiers

CAS 1119-97-7
Molecular Formula C17H38BrN
Molecular Weight (g/mol) 336.40
MDL Number MFCD00011770
InChI Key CXRFDZFCGOPDTD-UHFFFAOYSA-M
Synonym cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t
PubChem CID 14250
ChEBI CHEBI:3565
IUPAC Name trimethyl(tetradecyl)azanium bromide
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
pH 4.0 - 7.0 (50g/L)
Quantity 25 g
Formula Weight 336.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecyltrimethylammonium Bromide
EINECSNumber (2)-0182&(2)-0184&(9)-1971
RTECSNumber BS5776000
TSCA Yes