missing translation for 'onlineSavingsMsg'
Learn More

Tetradecylphosphonic Acid 98.0+%, TCI America™

$155.93 - $550.61

Chemical Identifiers

CAS 4671-75-4
Molecular Formula C14H31O3P
Molecular Weight (g/mol) 278.373
MDL Number MFCD00015834
InChI Key BVQJQTMSTANITJ-UHFFFAOYSA-N
Synonym phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate
PubChem CID 78401
IUPAC Name tetradecylphosphonic acid
SMILES CCCCCCCCCCCCCCP(=O)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T31831G
View Documents
TCI America
T31831G
1 g
Each for $155.93
Only null left
 
T31835G
View Documents
TCI America
T31835G
5 g
Each for $550.61
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4671-75-4
278.373
BVQJQTMSTANITJ-UHFFFAOYSA-N
78401
CCCCCCCCCCCCCCP(=O)(O)O
C14H31O3P
MFCD00015834
phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate
tetradecylphosphonic acid

Specifications

4671-75-4
White
MFCD00015834
phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate
CCCCCCCCCCCCCCP(=O)(O)O
278.373
278.37
Crystalline Powder
97°C
C14H31O3P
1 g
BVQJQTMSTANITJ-UHFFFAOYSA-N
tetradecylphosphonic acid
78401
≥98.0% (T)
Tetradecylphosphonic Acid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TA0460000

TSCA : Yes

SDS
Documents

Documents