accessibility menu, dialog, popup

UserName

H3CPOHOHO

CAS RN 4671-75-4

IUPAC Name: tetradecylphosphonic acid
Synonyms: phosphonic acid, tetradecyl-
Molecular Weight (g/mol): 278.37
Molecular Formula: C14H31O3P
InChi Key: BVQJQTMSTANITJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCP(O)(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)