missing translation for 'onlineSavingsMsg'
Learn More

Tetradecylphosphonic Acid 98.0+%, TCI America™

Catalog No. T31831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31831G 1 g
T31835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31831G Supplier TCI America Supplier No. T31831G

Chemical Identifiers

CAS 4671-75-4
Molecular Formula C14H31O3P
Molecular Weight (g/mol) 278.373
MDL Number MFCD00015834
InChI Key BVQJQTMSTANITJ-UHFFFAOYSA-N
Synonym phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate
PubChem CID 78401
IUPAC Name tetradecylphosphonic acid
SMILES CCCCCCCCCCCCCCP(=O)(O)O

Specifications

Melting Point 97°C
Color White
Quantity 1 g
Formula Weight 278.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecylphosphonic Acid
RTECSNumber TA0460000
TSCA Yes