missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™

Catalog No. p-7137339
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26535G 5 g
T265325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26535G Supplier TCI America Supplier No. T26535G
Only null left

Chemical Identifiers

CAS 14933-09-6
Molecular Formula C19H41NO3S
Molecular Weight (g/mol) 363.601
MDL Number MFCD00036910
InChI Key BHATUINFZWUDIX-UHFFFAOYSA-N
Synonym myristyl sulfobetaine, zwittergent 3-14, n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dimethyl tetradecyl ammonio propane-1-sulfonate, 3-n,n-dimethylmyristylammonio propanesulfonate, dimethyl 3-sulphonatopropyl tetradecylammonium, zwittergent™ 3-14 detergent, 3-myristyldimethylammonio propanesulfonate, 3-n,n-dimethyltetradecylammonio propanesulfonate
PubChem CID 84705
ChEBI CHEBI:10129
IUPAC Name 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Melting Point 245°C
Color White
Quantity 5 g
Formula Weight 363.60
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt [for Biochemical Research]