missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecylamine 96.0+%, TCI America™
Supplier: TCI America T009025G
Specifications
| Tetradecylamine | |
| White-Yellow | |
| C14H31N | |
| 25 g | |
| tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d | |
| CCCCCCCCCCCCCCN | |
| 213.409 | |
| 213.41 | |
| Crystalline Lumps |
| 2016-42-4 | |
| 291°C | |
| MFCD00008157 | |
| 3259 | |
| PLZVEHJLHYMBBY-UHFFFAOYSA-N | |
| tetradecan-1-amine | |
| 16217 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 2016-42-4 | |
| 213.409 | |
| PLZVEHJLHYMBBY-UHFFFAOYSA-N | |
| 16217 | |
| CCCCCCCCCCCCCCN |
| C14H31N | |
| MFCD00008157 | |
| tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d | |
| tetradecan-1-amine |
Safety and Handling
EINECSNumber : (2)-0133&(2)-0176
RTECSNumber : XB7970000
TSCA : Yes