missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecylamine 96.0+%, TCI America™
$72.70 - $247.92
Chemical Identifiers
| CAS | 2016-42-4 |
|---|---|
| Molecular Formula | C14H31N |
| Molecular Weight (g/mol) | 213.409 |
| MDL Number | MFCD00008157 |
| InChI Key | PLZVEHJLHYMBBY-UHFFFAOYSA-N |
| Synonym | tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d |
| PubChem CID | 16217 |
| IUPAC Name | tetradecan-1-amine |
| SMILES | CCCCCCCCCCCCCCN |
Chemical Identifiers
| 2016-42-4 | |
| 213.409 | |
| PLZVEHJLHYMBBY-UHFFFAOYSA-N | |
| 16217 | |
| CCCCCCCCCCCCCCN |
| C14H31N | |
| MFCD00008157 | |
| tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d | |
| tetradecan-1-amine |
Specifications
| 2016-42-4 | |
| 291°C | |
| MFCD00008157 | |
| 3259 | |
| PLZVEHJLHYMBBY-UHFFFAOYSA-N | |
| tetradecan-1-amine | |
| 16217 | |
| ≥96.0% (GC) | |
| Tetradecylamine |
| White-Yellow | |
| C14H31N | |
| 25 g | |
| tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d | |
| CCCCCCCCCCCCCCN | |
| 213.409 | |
| 213.41 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0133&(2)-0176
RTECSNumber : XB7970000
TSCA : Yes