missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyl Ether 97.0+%, TCI America™

Catalog No. t009125g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T009125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T009125G Supplier TCI America Supplier No. T009125G

Chemical Identifiers

CAS 5412-98-6
Molecular Formula C28H58O
Molecular Weight (g/mol) 410.771
MDL Number MFCD00026607
InChI Key HANWHVWXFQSQGJ-UHFFFAOYSA-N
Synonym Dimyristyl Ether, Ditetradecyl Ether, Myristyl Ether
PubChem CID 79433
IUPAC Name 1-tetradecoxytetradecane
SMILES CCCCCCCCCCCCCCOCCCCCCCCCCCCCC

Specifications

Melting Point 45°C
Color White
Quantity 25 g
Formula Weight 410.77
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecyl Ether