accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 5412-98-6

IUPAC Name: 1-(tetradecyloxy)tetradecane
Synonyms: dimyristyl ether
Molecular Weight (g/mol): 410.77
Molecular Formula: C28H58O
InChi Key: HANWHVWXFQSQGJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCOCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results