missing translation for 'onlineSavingsMsg'
Learn More

Tetradecenylsuccinic Anhydride 85.0+%, TCI America™

Catalog No. T0089500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0089500G 500 g
T008925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0089500G Supplier TCI America Supplier No. T0089500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33806-58-5
Molecular Formula C18H30O3
Molecular Weight (g/mol) 294.44
MDL Number MFCD00014543
InChI Key AXFMGCDLSGAENA-UHFFFAOYNA-N
PubChem CID 6043265
IUPAC Name 3-(tetradec-13-en-1-yl)oxolane-2,5-dione
SMILES C=CCCCCCCCCCCCCC1CC(=O)OC1=O

Specifications

Quantity 500 g
Formula Weight 294.44
Percent Purity ≥85.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecenylsuccinic Anhydride
EINECSNumber (2)-0852
TSCA Yes