accessibility menu, dialog, popup

UserName

CAS RN 33806-58-5

IUPAC Name: 3-(tetradec-13-en-1-yl)oxolane-2,5-dione
Synonyms: 3-(tetradec-13-en-1-yl)oxolane-2,5-dione
Molecular Weight (g/mol): 294.44
Molecular Formula: C18H30O3
InChi Key: AXFMGCDLSGAENA-UHFFFAOYNA-N
SMILES: C=CCCCCCCCCCCCCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results