missing translation for 'onlineSavingsMsg'
Learn More

Tetradecane, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014946
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014946 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014946 Supplier Merck Emd Millipore Supplier No. 871395ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetradecane is a hydrocarbon comprising of 14 carbon atoms and 30 hydrogen atoms. It can undergo combustion reaction to liberate carbon dioxide and water.

Specifications

CAS 629-59-4
Vapor Pressure 1 mmHg (76.4°C)
Vapor Density 6.83 (vs. air)
Density 0.762 g/mL (at 20°C)
Physical Form Neat
Linear Formula CH3(CH2)12 CH3
MDL Number MFCD00008986
Melting Point 5.5°C
Boiling Point 252°C to 254°C
Quantity 5 mL
Formula Weight 198.39
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less