missing translation for 'onlineSavingsMsg'
Learn More

Tetradecane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014945
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014945 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014945 Supplier Merck Emd Millipore Supplier No. 8713925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetradecane is a hydrocarbon comprising of 14 carbon atoms and 30 hydrogen atoms. It can undergo combustion reaction to liberate carbon dioxide and water.

Specifications

Description Olefine free
CAS 629-59-4
Ignition Point 455°F
Vapor Pressure 1 mmHg (76.4°C)
Vapor Density 6.83 (vs. air)
Density 0.762 g/mL (at 20°C)
Flash Point 102°C
Physical Form Neat
Linear Formula CH3(CH2)12 CH3
MDL Number MFCD00008986
Melting Point 5.5°C
Boiling Point 252°C to 254°C
Type Analytical Standard
Quantity 25 mL
Formula Weight 198.39
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less