missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America T252525G

 View more versions of this product

Catalog No. T252525G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetracycline Hydrochloride
214°C
C22H25ClN2O8
25 g
HTXDZWDXSWLLLW-FMZCEJRJSA-N
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
129628373
≥98.0% (T)
64-75-5
Yellow
MFCD00078142
tetracycline hydrochloride
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
480.898
480.90
Crystalline Powder

Chemical Identifiers

64-75-5
480.898
HTXDZWDXSWLLLW-FMZCEJRJSA-N
129628373
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
C22H25ClN2O8
MFCD00078142
tetracycline hydrochloride
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : QI9100000

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions