missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline Hydrochloride 98.0+%, TCI America™

Catalog No. p-7137262
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T252525G 25 g
T2525100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T252525G Supplier TCI America Supplier No. T252525G

Chemical Identifiers

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
MDL Number MFCD00078142
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
Synonym tetracycline hydrochloride
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Specifications

Melting Point 214°C
Color Yellow
Quantity 25 g
Formula Weight 480.90
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetracycline Hydrochloride
RTECSNumber QI9100000
TSCA Yes
Recommended Storage Freezer