missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracycline Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T2525100G
Specifications
| Tetracycline Hydrochloride | |
| 214°C | |
| C22H25ClN2O8 | |
| 100 g | |
| HTXDZWDXSWLLLW-FMZCEJRJSA-N | |
| (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride | |
| 129628373 | |
| ≥98.0% (T) |
| 64-75-5 | |
| Yellow | |
| MFCD00078142 | |
| tetracycline hydrochloride | |
| CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl | |
| 480.898 | |
| 480.90 | |
| Crystalline Powder |
Chemical Identifiers
| 64-75-5 | |
| 480.898 | |
| HTXDZWDXSWLLLW-FMZCEJRJSA-N | |
| 129628373 | |
| CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl |
| C22H25ClN2O8 | |
| MFCD00078142 | |
| tetracycline hydrochloride | |
| (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
Safety and Handling
RTECSNumber : QI9100000
TSCA : Yes
Recommended Storage : Freezer