Learn More
Tetracycline hydrochloride, 96%
Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria. | CAS: 64-75-5 | C22H25ClN2O8 | 480.898 g/mol
Supplier: Thermo Scientific Chemicals B2140822
Description
Tetracycline hydrochloride is used to induce apoptosis in osteoclasts. It is used to treat acne and other skin infections, respiratory tract infections like pneumonia, genital, urinary infections, leptospirosis, helicobacter pylori, taxoplasmosis, mycoplasma, psittacosis for dog and cats. It also works more effectively in animals that have tick-borne infections. It is also useful in cell culture applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Tetracycline hydrochloride is the hydrochloride salt of the antibiotic tetracycline
- Tetracycline blocks amino acyl-tRNA binding during bacterial protein synthesis
- Tetracycline is a broad-spectrum naphthacene antibiotic that inhibits growth of both gram positive and gram negative bacteria
Applications
- Tetracycline may be used as a selection agent in cell culture
- It may also be used to activate inducible gene expression in engineered cells
Specifications
| Tetracycline Hydrochloride | |
| 64-75-5 | |
| 100 g | |
| 0.98 | |
| C22H25ClN2O8 | |
| 14,9196 | |
| HTXDZWDXSWLLLW-FMZCEJRJSA-N | |
| (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride | |
| 129628373 | |
| 96% |
| Tetracycline hydrochloride | |
| 220°C to 223°C | |
| −247° (c=1, 0.1M HCl) | |
| Air and light sensitive | |
| MFCD00078142 | |
| Soluble in water,dimethyl sulfoxide,methanol and ethanol. Insoluble in ether and hydrocarbons. | |
| CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl | |
| 480.898 | |
| 480.91 |
Chemical Identifiers
| 64-75-5 | |
| 480.898 | |
| HTXDZWDXSWLLLW-FMZCEJRJSA-N | |
| (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
| C22H25ClN2O8 | |
| MFCD00078142 | |
| 129628373 | |
| CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl |
Safety and Handling
GHS H Statement
H361-H319
Suspected of damaging fertility or the unborn child.
Causes serious eye irritation.
P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
H319-H361d
EINECSNumber : 200-593-8
RTECSNumber : QI9100000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only