missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline hydrochloride, 96%

Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria. | CAS: 64-75-5 | C22H25ClN2O8 | 480.898 g/mol

Supplier:  Thermo Scientific Chemicals B2140814

 View more versions of this product

Catalog No. AAB2140814

Only null left
Explore available promotions

Description

Description

Tetracycline hydrochloride is used to induce apoptosis in osteoclasts. It is used to treat acne and other skin infections, respiratory tract infections like pneumonia, genital, urinary infections, leptospirosis, helicobacter pylori, taxoplasmosis, mycoplasma, psittacosis for dog and cats. It also works more effectively in animals that have tick-borne infections. It is also useful in cell culture applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tetracycline hydrochloride is the hydrochloride salt of the antibiotic tetracycline
  • Tetracycline blocks amino acyl-tRNA binding during bacterial protein synthesis
  • Tetracycline is a broad-spectrum naphthacene antibiotic that inhibits growth of both gram positive and gram negative bacteria

Applications

  • Tetracycline may be used as a selection agent in cell culture
  • It may also be used to activate inducible gene expression in engineered cells
Specifications

Specifications

Tetracycline Hydrochloride
64-75-5
25 g
Air and light sensitive
MFCD00078142
Soluble in water,dimethyl sulfoxide,methanol and ethanol. Insoluble in ether and hydrocarbons.
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
480.898
480.91
Tetracycline hydrochloride
220°C to 223°C
0.98
C22H25ClN2O8
14,9196
HTXDZWDXSWLLLW-FMZCEJRJSA-N
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
129628373
96%

Chemical Identifiers

64-75-5
480.898
HTXDZWDXSWLLLW-FMZCEJRJSA-N
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
C22H25ClN2O8
MFCD00078142
129628373
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H319

P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c

H319-H361d

EINECSNumber : 200-593-8

RTECSNumber : QI9100000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only