missing translation for 'onlineSavingsMsg'
Learn More

Tetracyanoethylene 98.0+%, TCI America™

Catalog No. p-7136067
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
T00775G 5 g
T007725G 25 g
2 options

Catalog No. T00775G

Supplier: TCI America T00775G

Only null left

Chemical Identifiers

CAS 670-54-2
Molecular Formula C6N4
Molecular Weight (g/mol) 128.094
MDL Number MFCD00001850
InChI Key NLDYACGHTUPAQU-UHFFFAOYSA-N
Synonym tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
PubChem CID 12635
IUPAC Name ethene-1,1,2,2-tetracarbonitrile
SMILES C(#N)C(=C(C#N)C#N)C#N

Specifications

Melting Point 198°C
Boiling Point 125°C
Quantity 5 g
UN Number 3439
Formula Weight 128.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetracyanoethylene
RTECSNumber KM7300000
TSCA Yes