missing translation for 'onlineSavingsMsg'
Learn More

Tetracyanoethylene (purified by sublimation) 98.0+%, TCI America™

Catalog No. T32641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32641G 1 g
T32645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32641G Supplier TCI America Supplier No. T32641G

Chemical Identifiers

CAS 670-54-2
Molecular Formula C6N4
Molecular Weight (g/mol) 128.094
MDL Number MFCD00001850
InChI Key NLDYACGHTUPAQU-UHFFFAOYSA-N
Synonym tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
PubChem CID 12635
IUPAC Name ethene-1,1,2,2-tetracarbonitrile
SMILES C(#N)C(=C(C#N)C#N)C#N

Specifications

Melting Point 199°C
Color White
Boiling Point 223°C
Quantity 1 g
UN Number 3439
Formula Weight 128.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetracyanoethylene (purified by sublimation)
RTECSNumber KM7300000
TSCA Yes