Learn More
Tetracyanoethylene, 98%
CAS: 670-54-2 | C6N4 | 128.094 g/mol
Supplier: Thermo Scientific Chemicals A1394506
Description
Used for postfunctional addition to polyphenylacetylene derivatives to change the oxygen permeability. As reactant for Regioselective [2+2] cycloaddition reaction for production of BODIPY dyes2 and TCBD derivatives, Thermal addition reaction with alkynes, One-pot reactions with nucleophilic reagents forming aromatic cyanovinyl compounds, Synthesis of cobalt tetracyanoethylene films, Biotransformation by Botrytis cinerea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed for postfunctional addition to polyphenylacetylene derivatives to change the oxygen permeability. As reactant for Regioselective [2+2] cycloaddition reaction for production of BODIPY dyes2 and TCBD derivatives, Thermal addition reaction with alkynes, One-pot reactions with nucleophilic reagents forming aromatic cyanovinyl compounds, Synthesis of cobalt tetracyanoethylene films, Biotransformation by Botrytis cinerea.
Solubility
Soluble in methanol, benzene, toluene, water (hydrolyses to form HCN), and ether (slightly).
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| Tetracyanoethylene | |
| 197°C to 199°C | |
| 223°C | |
| MFCD00001850 | |
| UN3439 | |
| Moisture sensitive | |
| tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene | |
| NLDYACGHTUPAQU-UHFFFAOYSA-N | |
| ethene-1,1,2,2-tetracarbonitrile | |
| 12635 | |
| 98% |
| 670-54-2 | |
| 1.348 | |
| C6N4 | |
| 5 g | |
| 1679885 | |
| 14,9195 | |
| Soluble in methanol,benzene,toluene,water (hydrolyses to form HCN),and ether (slightly). | |
| C(#N)C(=C(C#N)C#N)C#N | |
| 128.094 | |
| 128.09 |
Chemical Identifiers
| 670-54-2 | |
| 128.094 | |
| NLDYACGHTUPAQU-UHFFFAOYSA-N | |
| 12635 | |
| C(#N)C(=C(C#N)C#N)C#N |
| C6N4 | |
| MFCD00001850 | |
| tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene | |
| ethene-1,1,2,2-tetracarbonitrile |
Safety and Handling
GHS H Statement
H300-H310-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P363-P501c
H300+H310-H315-H319-H331-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: I
EINECSNumber : 211-578-0
RTECSNumber : KM7300000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only