missing translation for 'onlineSavingsMsg'
Learn More

Tetracosane, 99%

Catalog No. AC138040250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138040250 25 g Plastic bottle
AC138041000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC138040250 Supplier Thermo Scientific Chemicals Supplier No. 138040250

CAS: 646-31-1 | C24H50 | 338.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 646-31-1
Molecular Formula C24H50
Molecular Weight (g/mol) 338.65
MDL Number MFCD00009352
InChI Key POOSGDOYLQNASK-UHFFFAOYSA-N
Synonym n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b
PubChem CID 12592
ChEBI CHEBI:32936
IUPAC Name tetracosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 49°C to 52°C
Color White
Boiling Point 391°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3(CH2)22CH3
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 338.65
Percent Purity 99%
Physical Form Shiny Flakes
Chemical Name or Material Tetracosane, 99%
Show More Show Less
EINECSNumber 211-474-5
TSCA TSCA

RUO – Research Use Only