missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%

Catalog No. p-4408478
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA32437K2 1 L
AA32437M1 4 x 1 L
AA32437K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA32437K2 Supplier Thermo Scientific Chemicals Supplier No. 032437K2
Only null left

CAS: 127-18-4 | C2Cl4 | 165.82 g/mol

Tetrachloroethylene is used as solvent. It is involved in dry cleaning as well as in the degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name 1,1,2,2-tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point °C to 22°C
Density 1.623 g/mL
Boiling Point 120°C to 121°C
Assay Percent Range >99%
Refractive Index 1.5055
Quantity 1 L
Beilstein 1361721
Solubility Information Miscible with alcohol,ether,chloroform,benzene and hexane. Slightly miscible with water.
Formula Weight 165.83
Grade Spectrophotometric
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene, Ultrapure
Show More Show Less
Hazard Category H315-H317-H319-H336-H350-H373
Precautionary Statement P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-825-9
RTECSNumber KX3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only