missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haen
CHROMASOLV™, for HPLC,≥99.9%
$415.60 - $415.60
Chemical Identifiers
| CAS | 127-18-4 |
|---|---|
| Molecular Formula | C2Cl4 |
| Molecular Weight (g/mol) | 165.82 |
| MDL Number | MFCD00000834 |
| InChI Key | CYTYCFOTNPOANT-UHFFFAOYSA-N |
| Synonym | tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin |
| PubChem CID | 31373 |
| ChEBI | CHEBI:17300 |
| SMILES | ClC(Cl)=C(Cl)Cl |
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
| 127-18-4 | |
| 165.82 | |
| CYTYCFOTNPOANT-UHFFFAOYSA-N | |
| 31373 | |
| ClC(Cl)=C(Cl)Cl |
| C2Cl4 | |
| MFCD00000834 | |
| tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
| CHEBI:17300 |
Specifications
| 127-18-4 | |
| 100.0000% | |
| 1.623Â g/mLÂ at 25Â °C(lit.) | |
| Glass Bottle | |
| CCl2=CCl2 | |
| 1 L | |
| 1361721 | |
| CYTYCFOTNPOANT-UHFFFAOYSA-N | |
| 1,1,2,2-tetrachloroethene | |
| 31373 | |
| 165.83 | |
| HPLC |
| max. 1.0 at 290 nm, max. 0.03 at 400 nm, max. 0.05 at 350 nm, max. 0.10 at 305 nm, max. 0.20 at 300 nm, max. 0.30 at 295 nm | |
| -22°C (literature) | |
| 120°C to 122°C | |
| C2Cl4 | |
| MFCD00000834 | |
| UN1897 | |
| tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
| ClC(Cl)=C(Cl)Cl | |
| 165.82 | |
| CHEBI:17300 | |
| >99.9% | |
| Tetrachloroethylene, CHROMASOLV™, for HPLC, ≥99.9% |
Safety and Handling
ShelfLife : 900 days from date of manufacture
DOTInformation : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Tetrachloroethylene
EINECSNumber : 204-825-9
Recommended Storage : Room Temp