missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, 99+%, spectrophotometric grade

Catalog No. p-4510447
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167890010 1 L Glass bottle
AC167890025 2.5 L Glass bottle
AC167890100 10 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167890010 Supplier Thermo Scientific Chemicals Supplier No. 167890010
Only null left

CAS: 127-18-4 | C2Cl4 | 165.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Absorbance (1cm cell vs water), (1 cm cell vs water) at 0nm, 0.03 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 305nm, 0.3 max. at 295nm, 1 max. at 290nm
Decomposition Information >150°C
Melting Point -22.0°C
Density 1.6250g/mL
Boiling Point 120.0°C to 122.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C=CCl2
Refractive Index 1.505
Quantity 1 L
Residue after Evaporation 0.0005% max.
Beilstein 01, 187
Specific Gravity 1.625
Merck Index 15, 9335
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether, oils, chloroform, and benzene
IUPAC Name 1,1,2,2-tetrachloroethene
Viscosity 0.89 mPa.s (20°C)
Vapor Pressure 18mbar at 20°C
Formula Weight 165.83
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Causes serious eye irritati
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r
EINECSNumber 204-825-9

RUO – Research Use Only