missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, ≥99.5%, Honeywell™
≥99.5%
$958.68 - $958.68
Chemical Identifiers
| CAS | 127-18-4 |
|---|---|
| Molecular Formula | C2Cl4 |
| Molecular Weight (g/mol) | 165.82 |
| MDL Number | MFCD00000834 |
| InChI Key | CYTYCFOTNPOANT-UHFFFAOYSA-N |
| Synonym | tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin |
| PubChem CID | 31373 |
| ChEBI | CHEBI:17300 |
| IUPAC Name | tetrachloroethene |
| SMILES | ClC(Cl)=C(Cl)Cl |
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
| 127-18-4 | |
| 165.82 | |
| CYTYCFOTNPOANT-UHFFFAOYSA-N | |
| 31373 | |
| tetrachloroethene |
| C2Cl4 | |
| MFCD00000834 | |
| tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
| CHEBI:17300 | |
| ClC(Cl)=C(Cl)Cl |
Specifications
| 127-18-4 | |
| Colorless | |
| 121°C | |
| C2Cl4 | |
| CCl2=CCl2 | |
| 6 x 1 L | |
| 1361721 | |
| CYTYCFOTNPOANT-UHFFFAOYSA-N | |
| tetrachloroethene | |
| 5.83 (vs air) | |
| CHEBI:17300 | |
| 13mmHg (20°C), 19mmHg (25°C) | |
| Liquid |
| -22°C | |
| 1.623g/mL at 25°C | |
| Glass Bottle | |
| n20/D 1.505(lit.) | |
| MFCD00000834 | |
| UN1897 | |
| tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
| ClC(Cl)=C(Cl)Cl | |
| 165.82 | |
| 31373 | |
| 165.83g/mol | |
| ≥99.5% | |
| Tetrachloroethylene |
Safety and Handling
P280-P305 + P351 + P338
H315-H317-H319-H336-H351-H411
DOTInformation : 6.1/PGIII
EINECSNumber : 204-825-9
RTECSNumber : KX3850000