missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorobisphenol A 98.0+%, TCI America™

Catalog No. T006225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T006225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T006225G Supplier TCI America Supplier No. T006225G
Only null left

Chemical Identifiers

CAS 79-95-8
Molecular Formula C15H12Cl4O2
Molecular Weight (g/mol) 366.059
MDL Number MFCD00002178
InChI Key KYPYTERUKNKOLP-UHFFFAOYSA-N
Synonym 2,2-Bis(3,5-dichloro-4-hydroxyphenyl)propane, 4,4′C-Isopropylidenebis(2,6-dichlorophenol)
PubChem CID 6619
ChEBI CHEBI:35004
IUPAC Name 2,6-dichloro-4-[2-(3,5-dichloro-4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(C1=CC(=C(C(=C1)Cl)O)Cl)C2=CC(=C(C(=C2)Cl)O)Cl

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 366.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrachlorobisphenol A
EINECSNumber (4)-0204
RTECSNumber SL6250000
TSCA Yes