missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloro-o-benzoquinone, 97%

Catalog No. p-196391
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC137990050 5 g Glass bottle
AC137990250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC137990050 Supplier Thermo Scientific Chemicals Supplier No. 137990050

CAS: 2435-53-2 | C6Cl4O2 | 245.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2435-53-2
Molecular Formula C6Cl4O2
Molecular Weight (g/mol) 245.86
MDL Number MFCD00001646
InChI Key VRGCYEIGVVTZCC-UHFFFAOYSA-N
Synonym o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil
PubChem CID 73252
IUPAC Name 3,4,5,6-tetrachlorocyclohexa-3,5-diene-1,2-dione
SMILES ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl

Specifications

Melting Point 125.0°C to 129.0°C
Color Bordeaux
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6Cl4(=O)2
Quantity 5 g
Beilstein 07, 602
Fieser 01,128; 04,76; 05,104; 07,355; 09,102; 10,85
Solubility Information Solubility in water: insoluble. Other solubilities: 10g/L chloroform (20°C)
Formula Weight 245.88
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetrachloro-o-benzoquinone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 219-424-4
TSCA TSCA

RUO – Research Use Only