accessibility menu, dialog, popup

UserName

ClClOOClCl

CAS RN 2435-53-2

IUPAC Name: tetrachlorocyclohexa-3,5-diene-1,2-dione
Synonyms: tetrachlorocyclohexa-3,5-diene-1,2-dione
Molecular Weight (g/mol): 245.86
Molecular Formula: C6Cl4O2
InChi Key: VRGCYEIGVVTZCC-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)