missing translation for 'onlineSavingsMsg'
Learn More

Tetracarbonyldi-μ-chlorodirhodium(I), 97%

Catalog No. p-195878
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197602500 250 mg Glass bottle
AC197600010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197602500 Supplier Thermo Scientific Chemicals Supplier No. 197602500
Only null left

CAS: 14523-22-9 | C4Cl2O4Rh2 | 388.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14523-22-9
Molecular Formula C4Cl2O4Rh2
Molecular Weight (g/mol) 388.75
MDL Number MFCD00135610
InChI Key FGKDXBICTVUSPK-UHFFFAOYSA-L
Synonym tetracarbonyldi-mu-chlorodirhodium i, rhodium carbonyl chloride i, rhodium i carbonyl chloride, rhodium i dicarbonyl chloride dimer, di-mu-chloro-tetracarbonyldirhodium i, chlorodicarbonylrhodium i dimer, 4co.cl2rh2, bis chlorodicarbonyl rhodium, tetracarbonyldi-, i-chlorodirhodium i, tetracarbonyldi-micron-chlorodirhodium i
IUPAC Name bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Specifications

Melting Point 121.0°C
Color Brown to Red
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Rh)
Packaging Glass bottle
Linear Formula [Rh(CO)2Cl]2
Quantity 250 mg
UN Number 2811
Merck Index 15, 8312
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents (except, aliphatic hydrocarbons)
Formula Weight 388.76
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Tetracarbonyldi-µ-chlorodirhodium(I)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 238-540-6

RUO – Research Use Only