accessibility menu, dialog, popup

UserName

CAS RN 14523-22-9

IUPAC Name: bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride
Synonyms: bis(λ¹-rhodium(1+)) tetrakis(carbon monoxide) dichloride
Molecular Weight (g/mol): 388.75
Molecular Formula: C4Cl2O4Rh2
InChi Key: FGKDXBICTVUSPK-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)