missing translation for 'onlineSavingsMsg'
Learn More

Tetracaine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7136911
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16885G 5 g
T168825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16885G Supplier TCI America Supplier No. T16885G

Chemical Identifiers

CAS 136-47-0
Molecular Formula C15H25ClN2O2
Molecular Weight (g/mol) 300.827
MDL Number MFCD00038912
InChI Key PPWHTZKZQNXVAE-UHFFFAOYSA-N
Synonym tetracaine hydrochloride, tetracaine hcl, butethanol, butylocaine, anethain, anethaine, curtacain, decicain, decicaine, dessicain
PubChem CID 8695
IUPAC Name 2-(dimethylamino)ethyl 4-(butylamino)benzoate;hydrochloride
SMILES CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C.Cl

Specifications

Melting Point 149°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 300.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetracaine Hydrochloride
RTECSNumber DG4900000
TSCA No