missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylthiuram Disulfide 98.0+%, TCI America™

Catalog No. p-7136378
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T063525G 25 g
T0635500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T063525G Supplier TCI America Supplier No. T063525G

Chemical Identifiers

CAS 1634-02-2
Molecular Formula C18H36N2S4
Molecular Weight (g/mol) 408.74
MDL Number MFCD00027191
InChI Key PGAXJQVAHDTGBB-UHFFFAOYSA-N
Synonym tetrabutylthiuram disulfide, tetra-n-butylthiuram disulfide, tetrabutylthiuram disulphide, thiuram disulfide tetrabutyl, thiuram, tetrabutyl-, disulfide, tetrabutylthioperoxydicarbamic acid, disulfide, bis dibutylthiocarbamoyl, unii-d969886eab, bis dibutylthiocarbamoyl disulfide, thioperoxydicarbonic diamide h2n c s 2s2 , tetrabutyl
PubChem CID 15412
IUPAC Name dibutylcarbamothioylsulfanyl N,N-dibutylcarbamodithioate
SMILES CCCCN(CCCC)C(=S)SSC(=S)N(CCCC)CCCC

Specifications

Melting Point 33°C
Quantity 25 g
Formula Weight 408.74
Percent Purity ≥98.0% (N)
Physical Form Crystalline Lumps
Chemical Name or Material Tetrabutylthiuram Disulfide
RTECSNumber JO0800000
TSCA Yes