accessibility menu, dialog, popup

UserName

H3CNH3CSSSSNCH3CH3

CAS RN 1634-02-2

IUPAC Name: N,N-dibutyl[(dibutylcarbamothioyl)disulfanyl]carbothioamide
Synonyms: thioperoxydicarbonic diamide (((H2N)C(S))2S2), t
Molecular Weight (g/mol): 408.74
Molecular Formula: C18H36N2S4
InChi Key: PGAXJQVAHDTGBB-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C(=S)SSC(=S)N(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results