missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylphosphonium Hydroxide (40% in Water), TCI America™
Supplier: TCI America T2571250G
Specifications
| Tetrabutylphosphonium Hydroxide (40% in Water) | |
| Yellow | |
| MFCD00068456 | |
| 3267 | |
| DFQPZDGUFQJANM-UHFFFAOYSA-M | |
| tetrabutylphosphanium hydroxide | |
| 84487 | |
| Liquid |
| 14518-69-5 | |
| C16H37OP | |
| 250 g | |
| tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o | |
| [OH-].CCCC[P+](CCCC)(CCCC)CCCC | |
| 276.45 | |
| 276.44 |
Chemical Identifiers
| 14518-69-5 | |
| 276.45 | |
| DFQPZDGUFQJANM-UHFFFAOYSA-M | |
| 84487 | |
| [OH-].CCCC[P+](CCCC)(CCCC)CCCC |
| C16H37OP | |
| MFCD00068456 | |
| tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o | |
| tetrabutylphosphanium hydroxide |
Safety and Handling
TSCA : Yes