missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylphosphonium Hydroxide (40% in Water), TCI America™
$59.69 - $323.43
Chemical Identifiers
| CAS | 14518-69-5 |
|---|---|
| Molecular Formula | C16H37OP |
| Molecular Weight (g/mol) | 276.45 |
| MDL Number | MFCD00068456 |
| InChI Key | DFQPZDGUFQJANM-UHFFFAOYSA-M |
| Synonym | tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o |
| PubChem CID | 84487 |
| IUPAC Name | tetrabutylphosphanium hydroxide |
| SMILES | [OH-].CCCC[P+](CCCC)(CCCC)CCCC |
Chemical Identifiers
| 14518-69-5 | |
| 276.45 | |
| DFQPZDGUFQJANM-UHFFFAOYSA-M | |
| 84487 | |
| [OH-].CCCC[P+](CCCC)(CCCC)CCCC |
| C16H37OP | |
| MFCD00068456 | |
| tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o | |
| tetrabutylphosphanium hydroxide |
Specifications
| 14518-69-5 | |
| C16H37OP | |
| 25 g | |
| tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o | |
| [OH-].CCCC[P+](CCCC)(CCCC)CCCC | |
| 276.45 | |
| 276.44 | |
| Tetrabutylphosphonium Hydroxide (40% in Water) |
| Yellow | |
| MFCD00068456 | |
| 3267 | |
| DFQPZDGUFQJANM-UHFFFAOYSA-M | |
| tetrabutylphosphanium hydroxide | |
| 84487 | |
| Liquid |
Safety and Handling
TSCA : Yes