missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylphosphonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. T20075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20075G 5 g
T200725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20075G Supplier TCI America Supplier No. T20075G

Chemical Identifiers

CAS 111928-21-3
Molecular Formula C16H36F6P2
Molecular Weight (g/mol) 404.402
MDL Number MFCD01863114
InChI Key QLQAFNAPINCKBM-UHFFFAOYSA-N
PubChem CID 16218641
IUPAC Name tetrabutylphosphanium;hexafluorophosphate
SMILES CCCC[P+](CCCC)(CCCC)CCCC.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 217°C
Color White
Quantity 5 g
Formula Weight 404.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylphosphonium Hexafluorophosphate