accessibility menu, dialog, popup

UserName

H3CPCH3H3CH3CFPFFFFF

CAS RN 111928-21-3

IUPAC Name: hexafluoro-λ⁵-phosphanuide; tetrabutylphosphanium
Synonyms: tetrabutylphosphonium hexafluorophosphate
Molecular Weight (g/mol): 404.4
Molecular Formula: C16H36F6P2
InChi Key: QLQAFNAPINCKBM-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCC[P+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results