missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Triiodide 96.0+%, TCI America™

Supplier:  TCI America T127125G

 View more versions of this product

Catalog No. T127125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetrabutylammonium Triiodide
71°C
C48H108I3N3
25 g
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[I-].[I-].[I-]
1108.126
623.18
Crystalline Powder
13311-45-0
Black-Purple
MFCD00059123
MZYZRSVLEXUIRP-UHFFFAOYSA-K
tetrabutylazanium;triiodide
23500181
≥96.0% (T)

Chemical Identifiers

13311-45-0
1108.126
MZYZRSVLEXUIRP-UHFFFAOYSA-K
tetrabutylazanium;triiodide
C48H108I3N3
MFCD00059123
23500181
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[I-].[I-].[I-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

TSCA : No

SDS
Documents

Documents

Promotions

Promotions