missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Triiodide 96.0+%, TCI America™
$152.86 - $382.13
Chemical Identifiers
| CAS | 13311-45-0 |
|---|---|
| Molecular Formula | C48H108I3N3 |
| Molecular Weight (g/mol) | 1108.126 |
| MDL Number | MFCD00059123 |
| InChI Key | MZYZRSVLEXUIRP-UHFFFAOYSA-K |
| PubChem CID | 23500181 |
| IUPAC Name | tetrabutylazanium;triiodide |
| SMILES | CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[I-].[I-].[I-] |
Chemical Identifiers
| 13311-45-0 | |
| 1108.126 | |
| MZYZRSVLEXUIRP-UHFFFAOYSA-K | |
| tetrabutylazanium;triiodide |
| C48H108I3N3 | |
| MFCD00059123 | |
| 23500181 | |
| CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[I-].[I-].[I-] |
Specifications
| 13311-45-0 | |
| Black-Purple | |
| MFCD00059123 | |
| MZYZRSVLEXUIRP-UHFFFAOYSA-K | |
| tetrabutylazanium;triiodide | |
| 23500181 | |
| ≥96.0% (T) | |
| Tetrabutylammonium Triiodide |
| 71°C | |
| C48H108I3N3 | |
| 5 g | |
| CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[I-].[I-].[I-] | |
| 1108.126 | |
| 623.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : No