missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Tribromide 98.0+%, TCI America™

Catalog No. T1284500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1284500G 500 g
T128425G 25 g
T1284100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1284500G Supplier TCI America Supplier No. T1284500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38932-80-8
Molecular Formula C48H108Br3N3
Molecular Weight (g/mol) 967.125
MDL Number MFCD00012110
InChI Key SFLXUZPXEWWQNH-UHFFFAOYSA-K
Synonym tetrabutylammonium tribromide, tetra-n-butylammonium tribromide, tetrabutylammoniumtribromide, mono tetrabutylammonium tribromide, tetrabutylamoniumtribromide, acmc-1ct1n, tetrabutylamonium tribromide, tetrabutylazanium tribromide, tetrabutylammonium-tribromide, tetra-butylammonium tribromide
PubChem CID 23500184
IUPAC Name tetrabutylazanium;tribromide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-]

Specifications

Melting Point 73°C
Quantity 500 g
UN Number 1759
Formula Weight 482.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Tribromide
EINECSNumber (2)-0186
TSCA No