accessibility menu, dialog, popup

UserName

CAS RN 38932-80-8

IUPAC Name: tetrabutylazanium; tribroman-2-uide
Synonyms: tetrabutylammonium tribromide
Molecular Weight (g/mol): 482.18
Molecular Formula: C16H36Br3N
InChi Key: XXSLZJZUSYNITM-UHFFFAOYSA-N
SMILES: Br[Br-]Br.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (6)
Quantity 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)